EDTA, pH 8.0, 0.5M
À propos de cet article
Ethylenediaminetetraacetic acid is a polyamino carboxylic acid, commonly used to deactivate metal dependent enzymes, or as a chelating agent that binds to metal ions
Suitable for RNA storage. Offered in different molar concentration for all common biological applications.
3 Options Available Below
Product Details & Documents
Ethylenediaminetetraacetic acid is a polyamino carboxylic acid, commonly used to deactivate metal dependent enzymes, or as a chelating agent that binds to metal ions
Suitable for RNA storage. Offered in different molar concentration for all common biological applications.
Product Family Options
Informations sur le produit
- DésignationTailleType de conditionnementAvailabilityPrice
- 0.5M EDTA1 LPC bottleEach(1l)$261.75Case of 12 (1 l)$3,141.00Supplier Number: MRGF-1202
- 0.5M EDTA2 LPC bottleEach(1/Itm/Bx)$372.97Case of 6 (1/ Itm/Bx)$2,237.82Supplier Number: MRGF-1203
- 0.5M EDTA10 LPC bottleDiscontinued-Supplier Number: MRGF-1203-010L
- 0.5M EDTA10 LCubitainer®Log in pour la disponibilitéPallet of 20 (1/ Itm/Bx)Pallet of 20 (1/ Itm/Bx) - $25,823.20$25,823.20Each(1/Itm/Bx)$1,291.16Pallet of 20 (1/ Itm/Bx)$25,823.20Supplier Number: MRGF-1203-010Q
- 0.5M EDTA20 LCubitainer®Discontinued-Supplier Number: -
- 0.5M EDTA200 LPP bottleDiscontinued-Supplier Number: -