Vancomycin Disks, Hardy Diagnostics
75813-974
Product available on GSA Advantage®
:
- Description:Vancomycin Disks
- Size:50 disks
- Cat. no.:75813-974
- Supplier No.:Z7501
Vancomycin disks are used for the differentiation of anaerobic bacteria based on susceptibility to vancomycin.
- Differentiation of anaerobic bacteria
- Spring loaded for smooth and reliable dispensing
For the differentiation of anaerobic bacteria based on susceptibility to vancomycin. Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing from the single metal disk dispenser.