ContactUsLinkComponent
SabHI Agar with Blood, Chloramphenicol and Cycloheximide HardyFlask™, Hardy Diagnostics
Catalogue # 75813-970
Fournisseur:  Hardy Diagnostics
Product available on GSA Advantage®
Numéro CAS:  
undefined
SabHI Agar with Blood, Chloramphenicol and Cycloheximide HardyFlask™, Hardy Diagnostics
Catalogue # 75813-970
Fournisseur:  Hardy Diagnostics
Supplier Number:  X73
Product available on GSA Advantage®
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    SabHI Agar with Blood, Chloramphenicol and Cycloheximide HardyFlask™
  • Size:
    Pack of 20
  • Cat. no.:
    75813-970
  • Volume:
    12 ml
  • Supplier No.:
    X73
Spécifications
À propos de cet article
SabHI Agar with Blood, Chloramphenicol and Cycloheximide is used for the selective isolation of pathogenic fungi.
  • Selective isolation of pathogenic fungi
  • 5% sheep blood, chloramphenicol and cycloheximide
Sabouraud Dextrose and Brain Heart Infusion Agar with 5% sheep blood, chloramphenicol and cycloheximide, for the selective isolation of pathogenic fungi.