VWR® Rochester Ocshner Kocher Rat Tooth Forceps
Supplier: VWR International
Certificates
About this item
VWR® Rochester Ocshner Kocher Rat Tooth Forceps are constructed of surgical grade stainless steel
- Passivated
- Rockwell Hardness (HRC): 40–45
- Box-lock joint
- Surgical grade
- Compatible with Gamma/ETO/Autoclavable sterilization methods
These instruments are available in curved or straight formats, in a variety of lengths. All forceps feature serrated jaws with rat tooth tips in a 1 x 2 teeth configuration. Forceps are fully autoclavable, and can be sterilized by EtO/gamma radiation.
Frequently Bought Together