VWR® Bone Nipper
Catalog # 10806-284
Supplier: VWR International
Specifications
- Description:VWR® Bone Nipper
- Length:102 mm (4")
- Material:Stainless steel, surgical grade
- Cat. no.:10806-284
Specifications
About this item
The VWR® Bone Nipper is made from ASTM (899-94) Series 400 German stainless steel
- Passivated
- Surgical grade
- Rockwell Hardness (HRC): 50–55
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Designed for cutting cartilage and bone, this instrument features a strong, removable bar-spring that makes the shears self-opening.