VWR® Beyer Double-Action Bone Rongeur
Catalogue # 10806-262
Fournisseur: VWR International
Spécifications
- Description:VWR® Beyer Double-Action Bone Rongeur
- Length:178 mm (7")
- Material:Stainless steel, surgical grade
- Cat. no.:10806-262
Spécifications
À propos de cet article
The VWR® Beyer Double-Action Bone Rongeur is made from ASTM (899-94) Series 400 German stainless steel
- Passivated
- Surgical grade
- Rockwell Hardness (HRC): 50–55
- Compatible with Gamma/ETO/Autoclavable sterilization methods
The instrument is designed for cutting cartilage and bone, and features a strong, removable bar-spring that makes the shears self-opening.