VWR® Blumenthal Bone Rongeurs
Fournisseur: VWR International
Certificats
À propos de cet article
The VWR® Blumenthal Bone Rongeur is made from ASTM (899-94) Series 400 German stainless steel.
- Passivated
- Surgical grade
- Rockwell Hardness (HRC): 50–55
- Compatible with Gamma/ETO/Autoclavable sterilization methods
The instrument is designed for cutting cartilage and bone, and features a strong, removable bar-spring that makes these shears self-opening.
Frequently Bought Together