VWR® Dissecting Bone Cutting Forceps
VWR® Dissecting Bone Cutting Forceps, 127 mm (5"), Stainless steel, surgical grade
Catalog # 10806-298
Supplier: VWR International
Jump to:
Product Details & Documents
Surgical Grade Stainless Steel.
- Passivated
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Spécifications
- Désignation:VWR® Dissecting Bone Cutting Forceps
- Longueur:127 mm (5")
- Matière:Stainless steel, surgical grade