VWR® Nichrome/Aluminum Innoculating Loops
Supplier: VWR International
Certificates
About this item
VWR® Nichrome/Aluminum Innoculating Loops consist of a nichrome wire loop mounted to an aluminum handle.
- Passivated
- 203 mm (8") handles, available with insulation
- Compatible with Gamma/ETO/Autoclavable sterilization methods
The aluminum handle is available in either an insulated or non-insulated format. Innoculating loops are available in a variety of gauges, loop hole sizes, and wire types to accommodate an array of laboratory requirements.
Frequently Bought Together