Blood Agar
Supplier: Hardy Diagnostics
Certificates
About this item
Hardy Diagnostics Blood Agar products are recommended for use as general purpose growth media for the isolation, cultivation, and differentiation of a wide variety of microorganisms.
- Microorganism isolation, cultivation, differentiation
- General purpose growth media
Frequently Bought Together