VWR® Graefe Micro Hook
Supplier: VWR International
Jump to:
À propos de cet article
The VWR® Graefe Micro Hook is constructed from German steel.
- Passivated
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Devices are available in sharp or blunt formats.
2 Options Available Below
Product Details & Documents
The VWR® Graefe Micro Hook is constructed from German steel.
- Passivated
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Devices are available in sharp or blunt formats.
Product Family Options
Informations sur le produit
- DésignationMatièreLongueurTip StyleAvailabilityPrice
- VWR® Graefe Micro HookSteel11.4 cm (4.5")SharpLog in pour la disponibilitéPack of 12Pack of 12 - $1,044.01$1,044.01Prix promotionnel pour les nouveaux clientsEach$87.00Pack of 12$1,044.01
- VWR® Graefe Micro HookSteel11.4 cm (4.5")BluntLog in pour la disponibilitéPack of 12Pack of 12 - $1,044.01$1,044.01Prix promotionnel pour les nouveaux clientsEach$87.00Pack of 12$1,044.01