VWR® Bowman Micro Needle
The VWR® Bowman Micro Needle is constructed from German steel.
- Passivated
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Devices are available in straight or curved formats.
Frequently Bought Together
The VWR® Bowman Micro Needle is constructed from German steel.
Devices are available in straight or curved formats.