VWR® Nickel-Plated Steel Probe
10806-334
- Description:VWR® Nickel-Plated Steel Probe
- Length:15.2 cm (6")
- Material:Nickel-plated steel
- Curvature:Curved
- Cat. no.:10806-334
The VWR® Nickel-Plated Steel Probe is equipped with a round handle that is partly knurled to ensure a firm grip.
- Passivated
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Nickel-plated steel probe features a curved point, tapered and blunt at the end.