VWR® Spear Shaped Dissection Needle
Catalog # 10806-322
Supplier: VWR International
Specifications
- Description:VWR® Spear Shaped Dissection Needle, Single-Cutting Edge
- Length:5 cm (2")
- Material:Stainless steel
- Cat. no.:10806-322
- Tip Style:Half Spear Tips
Specifications
About this item
VWR® Spear Shaped Dissection Needles are made from ASTM (899-94) Series 400 Stainless Steel.
- Passivated
- Hexagonal
- Stainless steel handle
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Needles are available with either a single-cutting edge, or a double-cutting edge. Each format features a stainless steel handle.