VWR® Dissection Needle, Stainless Steel
These dissection needles are made from stainless steel and feature sharp points.
- Passivated
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Available in straight or bent formats. Supplied as needle only.
Frequently Bought Together