VWR® Allis Tissue Clamp
À propos de cet article
The VWR® Allis Tissue Clamp is a ratcheted forcep used for grasping organs and slippery or dense tissue during electrosurgery.
- Tip end varies depending on needs
- Ergonomic design
The tip end style varies to meet the needs of the procedure. The device is straight with a 4x5 teeth configuration that features inward curving blades.
2 Options Available Below
Product Details & Documents
The VWR® Allis Tissue Clamp is a ratcheted forcep used for grasping organs and slippery or dense tissue during electrosurgery.
- Tip end varies depending on needs
- Ergonomic design
The tip end style varies to meet the needs of the procedure. The device is straight with a 4x5 teeth configuration that features inward curving blades.
Product Family Options
Informations sur le produit
- DésignationMatièreLongueurJaw TypeCurvatureAvailabilityPrice
- VWR® Allis Tissue ClampStainless steel, surgical grade152 mm (6")4×5 teeth, inward curving bladesStraightLog in pour la disponibilitéPack of 12Pack of 12 - $522.46$522.46Prix promotionnel pour les nouveaux clientsEach$43.54Pack of 12$522.46
- VWR® Allis Tissue ClampStainless steel, surgical grade191 mm (7.5")4×5 teeth, inward curving bladesStraightLog in pour la disponibilitéPack of 12Pack of 12 - $692.55$692.55Prix promotionnel pour les nouveaux clientsEach$57.71Pack of 12$692.55