3-D Water Kit©
À propos de cet article
A fun tool to help teach water concepts.
- Ideal for grades 5-12
- Embedded magnets in atoms reflect the correct positive or negative charges
- Students experience molecular attraction or repulsion hands-on
Your students will use these magnetic water molecules to discover hydrogen bonding, make ice, dissolve salt, evaporate water, explore transpiration, create ethanol, and much more. Water Kits© enhance learning through hand-on activity and facilitate more in depth concept conversations. Your students can feel the various strengths of hydrogen, covalent and ionic bonds. They can explore the water cycle – even make ice. They also can show the differences between molecular adhesion, cohesion and capillary action and demonstrate surface tension, evaporation, condensation and solubility.
Materials Included (per cup):
1 Plastic cup
12 Water molecules
1 Sodium
1 Chloride
1 Ethane molecule
1 Hydroxyl group
1 six-cup tray (case of 8 only)
Materials Included (per cup):
1 Plastic cup
12 Water molecules
1 Sodium
1 Chloride
1 Ethane molecule
1 Hydroxyl group
1 six-cup tray (case of 8 only)
Ordering information: Assembly required. Includes assembly instructions and lesson resources.
Caution: WARNING: SMALL MAGNETS- This product contains small magnets. Swallowed magnets can stick together across intestines, causing serious infections and death. Seek immediate medical attention if magnets are swallowed or inhaled.
WARNING: CHOKING HAZARD - This product contains small parts and should be kept out of the reach of children under the age of 3, because the parts or the pieces may present a choking hazard to small children.
CAUTION: This is a science education product, not a toy. It is not intended for children under 8 years old.
3 Options Available Below
Product Details & Documents
A fun tool to help teach water concepts.
- Ideal for grades 5-12
- Embedded magnets in atoms reflect the correct positive or negative charges
- Students experience molecular attraction or repulsion hands-on
Your students will use these magnetic water molecules to discover hydrogen bonding, make ice, dissolve salt, evaporate water, explore transpiration, create ethanol, and much more. Water Kits© enhance learning through hand-on activity and facilitate more in depth concept conversations. Your students can feel the various strengths of hydrogen, covalent and ionic bonds. They can explore the water cycle – even make ice. They also can show the differences between molecular adhesion, cohesion and capillary action and demonstrate surface tension, evaporation, condensation and solubility.
Materials Included (per cup):
1 Plastic cup
12 Water molecules
1 Sodium
1 Chloride
1 Ethane molecule
1 Hydroxyl group
1 six-cup tray (case of 8 only)
Materials Included (per cup):
1 Plastic cup
12 Water molecules
1 Sodium
1 Chloride
1 Ethane molecule
1 Hydroxyl group
1 six-cup tray (case of 8 only)
Ordering information: Assembly required. Includes assembly instructions and lesson resources.
Caution: WARNING: SMALL MAGNETS- This product contains small magnets. Swallowed magnets can stick together across intestines, causing serious infections and death. Seek immediate medical attention if magnets are swallowed or inhaled.
WARNING: CHOKING HAZARD - This product contains small parts and should be kept out of the reach of children under the age of 3, because the parts or the pieces may present a choking hazard to small children.
CAUTION: This is a science education product, not a toy. It is not intended for children under 8 years old.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Water Kit© , single cupEach$127.80Case of 12$1,382.75Supplier Number: CSWK-01
- Water Kit© , Kit contains 6 cupsKit of 1$852.10Case of 8$6,136.25Supplier Number: CSWK-06
- Water Kit, single cupEach$1,893.69Case of 18$34,086.48Supplier Number: CSWK-01Spécifications:New ProductNew Product