innuMIX qPCR MasterMix Probe has been developed for fast, highly reproducible, real-time PCR and validated on the most common real-time instruments.
- Ready-to-use master mix for use in probe-based, real-time PCR
- Includes innuTaq Hot-A DNA Polymerase and high-quality dNTPs in a 2x formulation
- Simple, well-established handling combined with superior reproducibility
- Excellent PCR efficiency and slope
- Ideal for most commercially available real-time PCR instruments
innuMIX qPCR MasterMix Probe has been developed for fast, highly reproducible, real-time PCR and validated on the most common real-time instruments
The concentration of MgCl2 in the master mix is already ideal, eliminating the need to add any more.
The master mix can be readily combined with an extremely wide variety of probe systems, including TaqMan® and rehybridization probes. By delivering the perfect combination of the latest chemistry and PCR enhancer developments with a hot-start Taq DNA polymerase, this product allows researchers to achieve highly specific, ultrasensitive, real-time PCR results. Plus, it significantly reduces the time required to prepare the qPCR batch, while making the process radically easier to handle. Users now only need to add the template, probes and primers to the reaction, followed by enough water (suitable for real-time PCR) to produce the final volume prep work that can readily be carried out at room temperature.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Application:qPCR probe