To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Length:292 mm (11½")
- Material:Stainless steel
- Cat. no.:10183-852
- Supplier No.:90-8952
- Tip Type:Straight
Specifications
About this item
Sklar's Micro-Cut Kevorkian Biopsy Punch is commonly used during colposcopy procedures to excise tissue from the endocervix for biopsy
This product has a 3x8mm rectangular bite with 4 lower teeth. There is a basket to catch the tissue sample after excision. The Micro-Cut instrument features a knife honed edge on the upper jaw for a cleaner and more precise cut of the tissue. The spring style handle features one large ring that can fit multiple fingers and has a horn on the upper handle for thumb placement.