Barraquer-Dewecker Iris Scissors, OR Grade, Sklar
Product Details & Documents
Barraquer-Dewecker Iris Scissors are thumb-finger, hinge action scissors used for ophthalmic procedures
These fine, sharp scissors are ideal for detailed dissection of fine iris tissue.
Spécifications
- Longueur:57 mm (2¼")
- Matière:Stainless steel
- Curvature:Angled
- Blade Type:Blunt/Blunt, Smooth