KIMAX® Erlenmeyer Flasks with Screw Caps, Kimcote®
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Erlenmeyer flasks for preparing and storing media, and for use as shaker flasks.
- KimCote® plastic safety coating help prevent and contain spills in broken glassware
- Ultra-clear, extremely durable, and autoclavable
- Large openings and GL 45 threaded closures for easy access and tight sealing
- Marking spots and scales are durable white ceramic enamel
- Made from 33 expansion borosilicate glass
Delivery: Blue caps and clear pour rings included.
Frequently Bought Together