Plasmodesmata Slide
Plasmodesmata slide, Section, Iron Hematoxylin and Orange G
Catalog # 470181-364
Supplier: Avantor
Jump to:
Product Details & Documents
Pathway between plant cells.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Visible plasmodesmata extending across cell walls.
Spécifications
- Désignation:Plasmodesmata slide
- Stain:Iron Hematoxylin and Orange G
- Orientation:Section