ContactUsLinkComponent
BD BBL™ MycoPrep™ Specimen Digestion/Decontamination Kit, BD Diagnostics
Catalogue # CA-90003-160
Fournisseur:  BD
Numéro CAS:  
undefined
BD BBL™ MycoPrep™ Specimen Digestion/Decontamination Kit, BD Diagnostics
Catalogue # CA-90003-160
Fournisseur:  BD
Supplier Number:  240862
Numéro CAS:  
Spécifications
  • Description:
    BD BBL™ MycoPrep™ Specimen Digestion/Decontamination Kit, 10 75 mL bottles of NALC-NaOH Solution and 5 packages of Phosphate Buffer (pH 6.8)
  • Cat. no.:
    CA-90003-160
  • Supplier No.:
    240862
Spécifications
À propos de cet article
The BD BBL™ MycoPrep™ Specimen Digestion/Decontamination Kits are used for the digestion and decontamination of clinical specimens suspected to contain mycobacteria, especially Mycobacterium tuberculosis.
  • Gentle and effective digestion/decontamination of mycobacterial specimen
  • Saves materials, time and labor
  • Decreased safety hazards