Forensic Dental Analysis
Forensic Dental Analysis
Supplier:   Ward's Science
Forensic Dental Analysis
Supplier:  Ward's Science
Certificates
Forensic Dental Analysis
Forensic Dental Analysis
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Introduce Students to the Various Techniques Used in Forensic Dentistry
  • Eight activities in one kit
  • Students get to "solve a crime" while learning scientific techniques
Forensic dentistry is defined as the branch of forensic medicine that deals with identification through teeth, dental apparatus and their markings. The activities in this kit will introduce students to forensic dentistry while they try and identify the suspect in the case. The eight different activities in this kit include the concepts of creating a dental chart, creating a dental casting, creating a bite impression, identification of a patient from dental records, metric analysis of castings and metric analysis of bite marks.
Materials Included:
Dental Alginate
Nitrile Glove, Medium
Dental Impression Trays
Plaster of Paris
Plastic Cup
Measuring Cup
Plastic Cup
Metric Ruler
Plastic Scoop
Tongue Depressor
Toothpicks
Box Wax Bite Plates
Product Family Options
Informations sur le produit
  • Désignation
    Includes
    No. of Uses
    Availability
    Price
  • Forensic Dental Analysis
    Teacher ‘s Guide and Student Copymasters
    30
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $710.90
    $710.90
    Each
       $710.90
    Supplier Number: IS9021
  • Forensic Dental Analysis Refill
    30
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $472.40
    $472.40
    Each
       $472.40
    Supplier Number: IS9021-REFILL