Corning® Bar Coded External and Internal Threaded Polypropylene Cryogenic Vials, Corning
Supplier:   Corning
Corning® Bar Coded External and Internal Threaded Polypropylene Cryogenic Vials, Corning
Supplier:  Corning
Certificates
Corning® Bar Coded External and Internal Threaded Polypropylene Cryogenic Vials, Corning
Corning® Bar Coded External and Internal Threaded Polypropylene Cryogenic Vials, Corning
4 Options Available Below
Product Details & Documents
These PP cryogenic vials with PP cap protect valuable cells, biological solutions, and aqueous solutions in ultra-low temperature storage.
  • 2 ml capacity, round bottom vials
  • Unique 1D and 2D barcodes, and human readable characters
  • Manufactured from polypropylene to withstand temperatures to as low as –196 °C (–321 °F)
  • Caps designed for a secure seal, and are compatible with automatic decapping equipment
  • Locking base
  • Sterile to SAL 10⁻⁶
  • Certified nonpyrogenic and DNase/RNase-free
The accessory cyrogenic vial boxes are designed for use with 1D and 2D barcoded vials. The 81-Place box accommodates internal or external threaded vials. The 100-Place box accommodates internal threaded vials. Boxes are autoclavable.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Matière
    Avec bouchon
    Cap Material
    Sterility
    Availability
    Price
  • Cryogenic vial with internal thread, 2D barcode on bottom, 1D barcode on side
    2.00 ml
    PP
    Oui
    PE
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $765.85
    $765.85
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $765.85
    Supplier Number: 8670
  • Cryogenic vial with external thread, 2D barcode on bottom, 1D barcode on side
    2.00 ml
    PP
    Oui
    PE
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $765.85
    $765.85
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $765.85
    Supplier Number: 8671
  • Cryogenic vial with internal thread, 1D barcode on side
    2.00 ml
    PP
    Oui
    PE
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $504.90
    $504.90
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $504.90
    Supplier Number: 8672
  • Cryogenic vial with external thread, 1D barcode on side
    2.00 ml
    PP
    Oui
    PE
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $564.30
    $564.30
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $564.30
    Supplier Number: 8676
Cryogenic Storage Boxes
Informations sur le produit
  • Catalog #
    Désignation
    Matière
    Sterile
    Availability
    Price
  • 100-Place Cryogenic Vial Box
    Polycarbonate
    -
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $218.45
    $218.45
    Case of 10
       $218.45
Informations sur le produit
  • Catalog #
    Désignation
    Matière
    Sterile
    Availability
    Price
  • 81-Place Cryogenic Vial Box
    Polycarbonate
    -
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $217.06
    $217.06
    Case of 10
       $217.06