SP Bel-Art Plain Spinbar Magnetic Stirring Bars, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Smooth surface stirbars have no sharp or flat edges which maximizes contact with the stirring vessel.
- Durable
- Inert
- Autoclave safe
Alnico magnet encapsulated in FDA-grade, USP Class VI Teflon® PTFE low friction coating.
Certifications: Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Frequently Bought Together