Anti-Adra2a Rabbit Polyclonal Antibody
10168-848
: Genetex
- Antibody type:Primary
- Antigen name:Adrenergic Receptor Alpha 2A
- Antigen symbol:ADRA2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Size:100 μl
- Gene ID:11551
- Antigen synonyms:OTTHUMP00000058849|alpha-2A-adrenergic receptor|ALPHA2AAR|ADRAR|receptor |ZNF32 |alpha-2A-|alpha-2AAR subtype C10|adrenergic|ADRA2|ADRA2R|ADRA2A
- Storage buffer:PBS containing 1 mg/ml BSA and 0.05% sodium azide.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.6 mg/ml
- Immunogen:Synthetic peptide corressponding to residues R(218) I Y Q I A K R R T R V P P S R R G(235) of the 3rd intracellular loop of human A2AAR.
- Purification:Antigen affinity purification
- Cat. no.:10168-848
- Supplier No.:GTX82843
Rabbit polyclonal antibody to alpha 2a Adrenergic receptor
Type: Primary
Antigen: Adra2a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat