Anti-IgG Goat Polyclonal Antibody (DyLight® 680)
Catalogue # 10168-132
Fournisseur: Genetex
Spécifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:DyLight® 680
- Host:Goat
- Isotype:IgG
- Reactivity:Rabbit
- Size:500 μl
- Epitope:Heavy and Light Chain
- Storage temperature:Conjugate should be stable for at least one year at 4°C.
- Concentration:1 mg/ml
- Cat. no.:10168-132
- Supplier No.:GTX76793
Spécifications
À propos de cet article
DyLight680-conjugated Goat anti-Rabbit IgG Polyclonal antibody
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight680
Epitope: Heavy and Light Chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit