Anti-CALCOCO2 Mouse Monoclonal Antibody [clone: GT422]
10167-568
: Genetex
- Antibody type:Primary
- Antigen name:Calcium Binding And Coiled-coil Domain 2
- Antigen symbol:CALCOCO2
- Clonality:Monoclonal
- Clone:GT422
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:10241
- Antigen synonyms:Calcium binding and coiled-coil domain 2|NDP52
- Storage buffer:PBS
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human NDP52. The exact sequence is proprietary.
- Purification:Affinity purified by Protein G.
- Cat. no.:10167-568
- Supplier No.:GTX630396
Mouse Monoclonal antibody [GT422] to NDP52 (calcium binding and coiled-coil domain 2)
Type: Primary
Antigen: CALCOCO2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human