Anti-CD33 Mouse Monoclonal Antibody [clone: 2C6B7]
Catalogue # 10169-314
Fournisseur: Genetex
Spécifications
- Antibody type:Primary
- Antigen name:Cd33 Molecule
- Antigen symbol:CD33
- Clonality:Monoclonal
- Clone:2C6B7
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Size:100 μg
- Gene ID:945
- Antigen synonyms:SIGLEC-3|SIGLEC3|p67
- Storage buffer:Purified antibody in PBS with 0.05% sodium azide and 0.5% protein stabilizer.
- Storage temperature:4 °C; –20 °C for long term storage
- Concentration:1 mg/ml
- Immunogen:The immunogen corresponding to a region within amino acids 15 and 237 of human CD33 expressed in E. Coli.
- Cat. no.:10169-314
- Supplier No.:GTX60744
Spécifications
À propos de cet article
Mouse Monoclonal Antibody [2C6B7] to CD33
Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human