Anti-GNL3 Mouse Monoclonal Antibody [clone: 2C8D5]
10168-744
: Genetex
- Antibody type:Primary
- Antigen name:Guanine Nucleotide Binding Protein-Like 3 (Nucleolar)
- Antigen symbol:GNL3
- Clonality:Monoclonal
- Clone:2C8D5
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 μg
- Gene ID:26354
- Antigen synonyms:NNP47|NS|E2IG3|C77032
- Storage buffer:Purified antibody in PBS with 0.05% sodium azide and 0.5% protein stabilizer.
- Storage temperature:4 °C; –20 °C for long term storage
- Concentration:1 mg/ml
- Immunogen:The immunogen corresponding to a region within amino acids 1 and 226 of human GNL3 expressed in E. Coli.
- Cat. no.:10168-744
- Supplier No.:GTX60706
Mouse Monoclonal Antibody [2C8D5] to GNL3
Type: Primary
Antigen: GNL3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse