Order Entry
Canada
ContactUsLinkComponent
 
Anti-KCNQ1 Mouse Monoclonal Antibody [clone: 5E12]
  10168-954
 :  Genetex
Anti-KCNQ1 Mouse Monoclonal Antibody [clone: 5E12]
  10168-954
 :  Genetex
 :  GTX60528

 

  • Antibody type:
    Primary
  • Antigen name:
    Potassium Channel, Voltage Gated Kqt-Like Subfamily Q, Member 1
  • Antigen symbol:
    KCNQ1
  • Clonality:
    Monoclonal
  • Clone:
    5E12
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    3784
  • Antigen synonyms:
    Kv7.1|JLNS1|ATFB3|WRS|ATFB1|SQT2|LQT1|LQT|Kv1.9|KCNA8|KVLQT1|RWS|KCNA9
  • Storage buffer:
    Ascitic fluid containing 0.03% sodium azide.
  • Storage temperature:
    4 °C; –20 °C for long term storage
  • Immunogen:
    The immunogen corresponding to human KCNQ1 expressed in E. Coli.
  • Cat. no.:
    10168-954
  • Supplier No.:
    GTX60528

 

 

Mouse Monoclonal Antibody [5E12] to KCNQ1

Type: Primary
Antigen: KCNQ1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human