Anti-MYC Mouse Monoclonal Antibody [clone: 7E10]
Catalog # 10168-240
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:V-Myc Avian Myelocytomatosis Viral Oncogene Homolog
- Antigen symbol:MYC
- Clonality:Monoclonal
- Clone:7E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:4609
- Antigen synonyms:MRTL|bHLHe39|c-Myc|MYCC
- Storage buffer:Ascitic fluid containing 0.03% sodium azide.
- Storage temperature:4 °C; –20 °C for long term storage
- Immunogen:The immunogen corresponding to human MYC expressed in E. Coli.
- Cat. no.:10168-240
- Supplier No.:GTX60440
Specifications
About this item
Mouse monoclonal antibody [7E10] to c-Myc
Type: Primary
Antigen: MYC
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human