Anti-Csnk1g3 Rabbit Polyclonal Antibody
Catalog # 10164-124
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Casein kinase 1, gamma 3
- Antigen symbol:CSNK1G3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Size:400 μl
- Gene ID:70425
- Antigen synonyms:CSNK1G3L|CKI-gamma 3
- Storage buffer:PBS with 0.09% (W/V) sodium azide
- Storage temperature:Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles.
- Concentration:0.50 mg/ml
- Immunogen:This Mouse Csnk1g3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 333-361 amino acids from the C-terminal region of mouse Csnk1g3.
- Purification:Purified through a protein A column, followed by peptide affinity purification.
- Cat. no.:10164-124
- Supplier No.:GTX53489
Specifications
About this item
Rabbit polyclonal antibody to Casein Kinase I gamma 3
Type: Primary
Antigen: Csnk1g3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse