Anti-MUC12 Rabbit Polyclonal Antibody
10168-394
: Genetex
- Antibody type:Primary
- Antigen name:Mucin 12, Cell Surface Associated
- Antigen symbol:MUC12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 μg
- Gene ID:10071
- Antigen synonyms:MUC-12|MUC11|MUC-11
- Storage buffer:PBS with 0.02% sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Residues 541-555 [WEDQNLRESRFGLEN] of the human MUC 12 protein
- Purification:Antigen affinity purification
- Cat. no.:10168-394
- Supplier No.:GTX30149
Rabbit polyclonal to human MUC 12
Type: Primary
Antigen: MUC12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human