Anti-IgG Donkey Polyclonal Antibody (Cy5®)
10169-384
: Genetex
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Donkey
- Isotype:IgG
- Reactivity:Goat
- Size:100 μg
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 , 10 mg/mL Bovine Serum Albumin (BSA) , 0.01% (w/v) Sodium Azide
- Storage temperature:Keep as concentrated solution. For short-term storage, store at 4 °C. For long-term storage, aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles. Protect from light.
- Concentration:1 mg/ml
- Immunogen:Goat IgG whole molecule
- Purification:Immunogen affinity purified
- Cat. no.:10169-384
- Supplier No.:GTX26566
Cy5-conjugated Donkey anti-Goat IgG Polyclonal antibody, pre-adsorbed
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Goat