Anti-Irs1 Rabbit Polyclonal Antibody
Catalogue # 10168-802
Fournisseur: Genetex
Spécifications
- Antibody type:Primary
- Antigen name:Insulin Receptor Substrate 1
- Antigen symbol:IRS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Size:100 μg
- Gene ID:16367
- Antigen synonyms:IRS 1|HIRS 1|IRS1|HIRS1|IRS-1|IRS1Tyr1229|Insulin Receptor Substrate 1|p-IRS1
- Storage buffer:PBS containing 1 mg/ml BSA and 0.05% sodium azide.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Immunogen:Synthetic peptide corresponding to residues D(631) Y M P M S P K S V S A P Q Q I(646) of human IRS-1.
- Purification:Antigen affinity purification
- Cat. no.:10168-802
- Supplier No.:GTX25604
Spécifications
À propos de cet article
Rabbit Polyclonal antibody to IRS1
Type: Primary
Antigen: Irs1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse