Anti-CRY1 Rabbit Polyclonal Antibody
Catalog # 10168-700
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Cryptochrome Circadian Clock 1
- Antigen symbol:CRY1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Size:100 μg
- Gene ID:1407
- Antigen synonyms:PHLL1
- Storage buffer:PBS containing 1 mg/ml BSA and 0.05% sodium azide.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic Peptide: C Q(594) S V G P K V Q R Q S S N(606)
- Purification:Antigen affinity purification
- Cat. no.:10168-700
- Supplier No.:GTX23518
Specifications
About this item
Rabbit Polyclonal antibody to Cryptochrome 1
Type: Primary
Antigen: CRY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse