Anti-CAPNS1 Mouse Monoclonal Antibody [clone: 156]
Catalog # 10168-694
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Calpain (28 kDa Subunit)
- Antigen symbol:CAPNS1
- Clonality:Monoclonal
- Clone:156
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Bovine
- Size:100 μl
- Gene ID:281664
- Storage buffer:Ascites containing 0.05 % sodium azide
- Storage temperature:–20 °C, avoid freeze/thaw cycles
- Concentration:6.80 mg/ml
- Immunogen:Purified bovine skeletal muscle 28 kDa calpain subunit.
- Cat. no.:10168-694
- Supplier No.:GTX23514
Specifications
About this item
Mouse monoclonal antibody [156] to Calpain S1
Type: Primary
Antigen: CAPNS1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine