Anti-VAMP4 Rabbit Polyclonal Antibody
Catalog # 10168-610
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Vesicle-Associated Membrane Protein 4
- Antigen symbol:VAMP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Size:100 μg
- Gene ID:8674
- Antigen synonyms:VAMP24|VAMP-4
- Storage buffer:PBS containing 1 mg/ml BSA and 0.05% sodium azide.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant rat VAMP-4 protein.
- Purification:Antigen affinity purification
- Cat. no.:10168-610
- Supplier No.:GTX23348
Specifications
About this item
Rabbit Polyclonal antibody to VAMP4
Type: Primary
Antigen: VAMP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat