Anti-Dlg2 Rabbit Polyclonal Antibody
Catalogue # 10168-584
Fournisseur: Genetex
Spécifications
- Antibody type:Primary
- Antigen name:Discs, Large Homolog 2 (Drosophila)
- Antigen symbol:DLG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:100 μg
- Gene ID:64053
- Antigen synonyms:Postsynaptic density protein PSD-93Channel-associated protein of synapse-110|PPP1R58|PSD93,Chapsyn-110|Chapsyn-1|PSD-93|Post Synaptic Density 93|Discs large homolog 2
- Storage buffer:PBS containing 1 mg/ml BSA and 0.05% sodium azide.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide corresponding to residues N(352) K L C D K P A S P R H Y S P(366) of human PSD-93.
- Purification:Antigen affinity purification
- Cat. no.:10168-584
- Supplier No.:GTX22930
Spécifications
À propos de cet article
Rabbit Polyclonal antibody to PSD93
Type: Primary
Antigen: Dlg2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat