Order Entry
Canada
ContactUsLinkComponent
 
Anti-PSEN1 Mouse Monoclonal Antibody [clone: APS 18]
  10167-916
 :  Genetex
Anti-PSEN1 Mouse Monoclonal Antibody [clone: APS 18]
  10167-916
 :  Genetex
 :  GTX15458
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody type:
    Primary
  • Antigen name:
    Presenilin 1
  • Antigen symbol:
    PSEN1
  • Clonality:
    Monoclonal
  • Clone:
    APS 18
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Primates,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    200 μg
  • Gene ID:
    5663
  • Antigen synonyms:
    FAD|Alzheimer Disease 3|Homo Sapiens Clone CC44 Senilin 1|Senilin 1|PS1|PSNL1|AD3|S182 Protein
  • Storage buffer:
    PBS containing 0.05 % sodium azide and 1 mg/ml BSA
  • Storage temperature:
    –20 °C, avoid freeze/thaw cycles
  • Concentration:
    1 mg/ml
  • Immunogen:
    Synthetic peptide corresponding to residues C S(313) K Y N A E S T E R E S Q D T V A E N D D G(334) of humen PS1.
  • Purification:
    Protein G
  • Cat. no.:
    10167-916
  • Supplier No.:
    GTX15458

 

 

Mouse monoclonal antibody [APS 18] to Presenilin-1

Type: Primary
Antigen: PSEN1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Primates