Anti-HMGB2 Rabbit Polyclonal Antibody
10166-820
: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:High Mobility Group Box 2
- Antigen symbol:HMGB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:3148
- Antigen synonyms:HMG2
- Storage buffer:1XPBS, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human HMGB2. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:10166-820
- Supplier No.:GTX129464
Rabbit Polyclonal antibody to HMGB2 (high mobility group box 2)
Type: Primary
Antigen: HMGB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human