Order Entry
Canada
ContactUsLinkComponent
 
Anti-SQSTM1 Rabbit Polyclonal Antibody
  10166-774
 :  Genetex
Anti-SQSTM1 Rabbit Polyclonal Antibody
  10166-774
 :  Genetex
 :  GTX128173
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody type:
    Primary
  • Antigen name:
    Sequestosome 1
  • Antigen symbol:
    SQSTM1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    8878
  • Antigen synonyms:
    OSIL|A170|FTDALS3|ZIP3|p60|p62B|p62|PDB3
  • Storage buffer:
    1XPBS, 1% BSA, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SQSTM1. The exact sequence is proprietary.
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. no.:
    10166-774
  • Supplier No.:
    GTX128173

 

 

Rabbit Polyclonal antibody to SQSTM1 (sequestosome 1)

Type: Primary
Antigen: SQSTM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human