Order Entry
Canada
ContactUsLinkComponent
Anti-NFIA Rabbit Polyclonal Antibody
  10166-616
 :  Genetex
Anti-NFIA Rabbit Polyclonal Antibody
  10166-616
 :  Genetex
 :  GTX114580
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody type:
    Primary
  • Antigen name:
    Nuclear Factor I/A
  • Antigen symbol:
    NFIA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    4774
  • Antigen synonyms:
    NFI-A|NF1-A|Nuclear factor 1 A type|FLJ39164|Nuclear factor I/X|Nuclear factor 1 A-type|CCAAT Box Binding Transcription Factor|NFI L|Nuclear factor I/A|ynuclear factor I/X CCAAT-binding transcription factor|Nuclear factor 1/A|CTF|NF1 A|nuclear factor I/X CCAAT-binding transcription factor|Nuclear Factor I A|TGGCA-binding protein.|NFIA|CCAAT binding transcription factor|CCAAT-box-binding transcription factor|Nuclear Factor 1A|Nuclear Factor IA|NF I/A|NFI A|NF1A|TGGCA binding protein|NFIA_HUMAN|NF-I/A|Nuclear Factor 1 A
  • Storage buffer:
    1XPBS, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human NFIA. The exact sequence is proprietary.
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. no.:
    10166-616
  • Supplier No.:
    GTX114580

 

 

Rabbit Polyclonal antibody to NFIA (nuclear factor I/A)

Type: Primary
Antigen: NFIA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat