Anti-ABI1 Rabbit Polyclonal Antibody
Catalog # 89424-160
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Abl-Interactor 1
- Antigen symbol:ABI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 μl
- Gene ID:10006
- Antigen synonyms:Spectrin SH3 domain-binding protein 1|Abl-binding protein 4|SSH3BP|Eps8 SH3 domain-binding protein|Abl interactor 1|Nap1-binding protein|E3B1|Abelson interactor 1|SSH3BP1|ABLBP4|NAP1BP|ABI-1
- Storage buffer:1XPBS, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human SSH3BP1. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89424-160
- Supplier No.:GTX111478
Specifications
About this item
Rabbit Polyclonal antibody to SSH3BP1 (abl-interactor 1)
Type: Primary
Antigen: ABI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat