Order Entry
Canada
ContactUsLinkComponent
Anti-GTPBP4 Rabbit Polyclonal Antibody
Catalogue # 10166-562
Fournisseur:  Genetex
Anti-GTPBP4 Rabbit Polyclonal Antibody
Catalogue # 10166-562
Fournisseur:  Genetex
Supplier Number:  GTX110825
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    Gtp Binding Protein 4
  • Antigen symbol:
    GTPBP4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    23560
  • Antigen synonyms:
    NGB|NOG1|CRFG
  • Storage buffer:
    1XPBS, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Storage temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human GTPBP4. The exact sequence is proprietary.
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. no.:
    10166-562
  • Supplier No.:
    GTX110825

Spécifications

À propos de cet article

Rabbit Polyclonal antibody to GTPBP4 (GTP binding protein 4)

Type: Primary
Antigen: GTPBP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat