Anti-TYMS Rabbit Polyclonal Antibody
Catalogue # 89424-054
Fournisseur: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Thymidylate Synthase
- Antigen symbol:TYMS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 μl
- Gene ID:7298
- Antigen synonyms:TS|d TMP synthase|EC 2.1.1.45|TMS|MGC88736|HsT422
- Storage buffer:1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human Thymidylate synthetase. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89424-054
- Supplier No.:GTX103235
Spécifications
À propos de cet article
Rabbit Polyclonal antibody to Thymidylate synthetase (thymidylate synthetase)
Type: Primary
Antigen: TYMS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat