Accessory Parts for TRIO.BAS™ Air Samplers
An assortment of accessory and replacement products designed for use with the Hardy Diagnostics collection of TRIO.BAS™ air samplers.
Aspirating heads are available in stainless steel, Daily Shift polystyrene, or polycarbonate formats. The durable stainless steel and polycarbonate models are autoclavable and can be sprayed with disinfectant. The Daily Shift polystyrene models are supplied ready to use in a cleanroom environment. These heads are individually packaged with a Certificate of Sterility provided. The Blind Head is designed to protect the aspirating chamber when instrument is not in use.
: Stainless steel aspirating head models BAS330 and BAS331 and induction battery charger model BAS318 are required when instruments are purchased a la carte instead of the kit format. Vertical hook and wall holder require knob (sold separately).
Required Accessories
Frequently Bought Together