Trio.Bas™ MONO Kits
Supplier: Hardy Diagnostics
Certificates
About this item
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.
- Aspirating head with easy manipulation bayonet closure
- Manual or automatic operations
- Ergonomic and balanced design to facilitate handling with ease
- No plugs or external connections
All of the air samplers are lightweight, with an antibacterial techno-polymer shockproof body. They have a long lasting battery and are IP65 protected from dust and water
Per USP 797, 'Impaction is the preferred method of active air sampling. Use of settling plates for qualitative air sampling cannot be relied upon and shall not be used solely to determine the quality of air in the controlled environment.'
Certifications: ISO14698, USP 797, USP 116, EU GPL-GMP, ISO 17025.
Accessories information: Additional accessory components, including data transfer options, are available separately.
Delivery: Trio.Bas™ Mono kit contains:
Trio.Bas™ Mono air sampler with Bluetooth data transfer, includes calibration certificate
Induction battery charger, unit simply rests on base station to charge
Stainless steel aspirating head, autoclavable
Stainless steel cover head, autoclavable
Robustus hard shell carrying case to protect sampler in transit
IQ, OQ, PQ, filllable documents
Trio.Bas™ Mono air sampler with Bluetooth data transfer, includes calibration certificate
Induction battery charger, unit simply rests on base station to charge
Stainless steel aspirating head, autoclavable
Stainless steel cover head, autoclavable
Robustus hard shell carrying case to protect sampler in transit
IQ, OQ, PQ, filllable documents